Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EMSURE® Sodium hydroxide pellets for analysis

Supplier: MilliporeSigma
Danger
Synonyms: Caustic sodaLyeSoda lyeSodium hydrate

EMSURE®
1.06498.5000 1.06498.9025 1.06498.0500 1.06498.9050


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Specification Test Results
Solubility 1090 g/l (20 °C)
Melting Point 323 °C
Molar Mass 40.00 g/mol
Boiling Point 1390 °C (1013 hPa)
Vapor Pressure (20 °C)
Density 2.13 g/cm3 (20 °C)
pH 14 (50 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
500 g Plastic Bottle 1.06498.0500 EM1.06498.0500 Case of 6 (500g) Retrieving Restricted
Each (500g) Retrieving Restricted
5 kg Plastic Bottle 1.06498.5000 EM1.06498.5000 Case of 4 (5kg) Retrieving Restricted
Each (5kg) Retrieving Restricted
25 kg Plastic Bottle 1.06498.9025 EM1.06498.9025 Each (25kg) Retrieving Restricted
50 kg Plastic Bottle 1.06498.9050 EM1.06498.9050 Each (50kg) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping