Sodium metaarsenite 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ100
1.06277.1000
1.06277.1000
EM1.06277.1000CS
1224
USD
CA1.06277.1000
EM1.06277.1000
Sodium metaarsenite 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®
Sodium metaarsenite
Formula:
NaAsO₂ MW: 129.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003472 CAS Number: 7784-46-5 EINECS: 232-070-5 UN: 1686 ADR: 6.1,II |
Specification Test Results
Form | liquid |
Amount-of-substance concentration | 0.04975 - 0.05025 |
Measurement uncertainty | +/- 0.00015 |
Traceability | NIST SRM |
The concentration is determined by volumetric titration and refers to 20°C. | Toxic Materials |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...