α-Naphtholphthalein indicator, Supelco®
Supplier: MilliporeSigma
Synonyms:
3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
1.06246.0001
1.06246.0005
1.06246.0005
1.06246.0001
EM1.06246.0001CS
25992
USD
CA1.06246.0005
EM1.06246.0001
EM1.06246.0005
CA1.06246.0001
α-Naphtholphthalein indicator, Supelco®
α-Naphtholphthalein
Formula:
C₂₈H₁₈O₄ MW: 418.45 g/mol |
MDL Number:
MFCD00036202 CAS Number: 596-01-0 EINECS: 209-875-5 |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | 253 - 255 °C |
Molar Mass | 418.45 g/mol |
Bulk Density | 350 kg/m3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...