1-Naphtholbenzeine indicator, Millipore®

Supplier: MilliporeSigma

Synonyms: alpha-Naphtholbenzeinα-Naphtholbenzein4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanolα-Naphtholbenzeinep-Naphtholbenzeine1-NaphtholbenzeinBis(4-hydroxy-1-naphthyl)phenylmethanolp-Naphtholbenzein

1.06202.0005 1.06202.0005
EM1.06202.0005CS 7567 USD
CA1.06202.0005 EM1.06202.0005
1-Naphtholbenzeine indicator, Millipore®
1-Naphtholbenzeine
Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
EINECS: 205-656-3

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 243 - 245 °C
Molar Mass 374.44 g/mol
Bulk Density 120 - 300 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR