Lithium carbonate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.05680.0250
1.05680.0250
EM1.05680.0250CS
4840
USD
CA1.05680.0250
EM1.05680.0250
Lithium carbonate, EMSURE® for analysis, Supelco®
Lithium carbonate
1.05680.0250 replaces and is the same as LX0325-1. Different Pack Size, Same Specifications.
1.05680.0250 is offered as an alternative to LX0325-20. different size, different packaging, same chem spec.
Formula:
Li₂CO₃ MW: 73.89 g/mol Melting Pt: 720 °C Density: 2.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 13,05549 |
Specification Test Results
Solubility | 13 g/l (20 °C) |
Melting Point | 720 °C |
Molar Mass | 73.89 g/mol |
Bulk Density | 250 kg/m3 |
Vapor Pressure | (20 °C) |
Density | 2.1 g/cm3 (20 °C) |
pH | 10 - 11 (5 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...