m-Cresol purple indicator, Supelco®

Supplier: MilliporeSigma

Synonyms: m-Cresolsulfonphthalein2',2''-Dimethylphenolsulphonphthalein3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxideMCP2',2''-Dimethylphenolsulfonphthaleinm-Cresolsulphonphthalein

1.05228.0005 1.05228.0005
EM1.05228.0005CS 11575 USD
CA1.05228.0005 EM1.05228.0005
m-Cresol purple indicator, Supelco®
m-Cresol purple
Formula: C₂₁H₁₈O₅S
MW: 382.43 g/mol
MDL Number: MFCD00005871
CAS Number: 2303-01-7
EINECS: 218-960-6

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Molar Mass 382.43 g/mol
Bulk Density 480 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR