Potassium nitrite, crystals, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.05067.0250
1.05067.0250
EM1.05067.0250CS
7829
USD
CA1.05067.0250
EM1.05067.0250
Potassium nitrite, crystals, EMSURE® for analysis, Supelco®
Potassium nitrite
Formula:
KNO₂ MW: 85.1 g/mol Melting Pt: 387 °C Density: 1.92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011408 CAS Number: 7758-09-0 EINECS: 231-832-4 UN: 1488 ADR: 5.1,II Merck Index: 12,07816 |
Specification Test Results
Solubility | 2810 g/l (20 °C) |
Melting Point | 440 °C |
Molar Mass | 85.11 g/mol |
Bulk Density | 980 kg/m3 |
Density | 1.92 g/cm3 (20 °C) |
pH | 7 - 10 (50 g/l, H2O, 20 °C) |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Hazard Class Description | Oxidizer |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...