Potassium hydrogen diiodate, EMSURE® for analysis, Supelco®

Danger

Synonyms: Potassium bi-iodate

1.04867.0050 1.04867.0050
EM1.04867.0050CS 21236 USD
EM1.04867.0050 CA1.04867.0050
Potassium hydrogen diiodate, EMSURE® for analysis, Supelco®
Potassium hydrogen diiodate
Formula: HI₂KO₆
MW: 389.91
MDL Number: MFCD00011400
CAS Number: 13455-24-8
EINECS: 236-650-9
UN: 3085
ADR: 5.1 (8),II

Order Now

Specification Test Results

Solubility 13 g/l (15 °C)
Molar Mass 389.91 g/mol
Bulk Density 1750 kg/m3
pH 1 - 2 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR