Potassium hydrogen carbonate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Potassium bicarbonate

1.04854.0500 1.04854.0500
EM1.04854.0500CS 1105 USD
CA1.04854.0500 EM1.04854.0500
Potassium hydrogen carbonate, EMSURE® for analysis, Supelco®
Potassium hydrogen carbonate

1.04854.0500 replaces and is the same as PX1345-1. Part # Change only, Same Pack Size, Same Specifications.

Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 12,07770

Order Now

Specification Test Results

Solubility 333 g/l (20 °C)
Molar Mass 100.12 g/mol
Bulk Density 900 - 1100 kg/m3
Density 2.17 g/cm3 (20 °C)
pH 8.0 - 8.6 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR