Sodium diphenylamine-4-sulfonate redox indicator, Millipore®

Supplier: MilliporeSigma

Synonyms: Sodium N-phenylsulphanilateSodium (4-anilinobenzenesulphonate)Sodium DiphenylaminesulphonateDiphenylamine-4-sulphonic acid, sodium salt

1.03590.0010 1.03590.0010
EM1.03590.0010CS 72994 USD
EM1.03590.0010 CA1.03590.0010
Sodium diphenylamine-4-sulfonate redox indicator, Millipore®
Sodium diphenylamine-4-sulphonate
Formula: C₁₂H₁₀NNaO₃S
MW: 271.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Order Now

Specification Test Results

Solubility 820 g/l (20 °C)
Melting Point 200 °C
Molar Mass 271.26 g/mol
Bulk Density 160 kg/m3
pH 5.7 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR