Calcium hydroxide for the preparation of DIN 19266 pH standard buffer solutions, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Slaked lime
1.02111.0025
1.02111.0025
EM1.02111.0025CS
16669
USD
EM1.02111.0025
CA1.02111.0025
Calcium hydroxide for the preparation of DIN 19266 pH standard buffer solutions, Supelco®
Calcium hydroxide
Formula:
Ca(OH)₂ MW: 74.09 g/mol Melting Pt: 550 °C Density: 2.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 EINECS: 215-137-3 ADR: 8,III Merck Index: 13,01675 |
Specification Test Results
Solubility | 1.7 g/l (20 °C) |
Melting Point | 550 °C (decomposition) |
Molar Mass | 74.09 g/mol |
Bulk Density | 400 kg/m3 |
Boiling Point | 2850 °C (1013 hPa) (decomposition) |
Vapor Pressure | (20 °C) |
Density | 2.24 g/cm3 (20 °C) |
pH | 12.6 (H2O, 20 °C) (saturated solution) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...