Ammonium carbamate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Carbamic acid ammonium salt

1.01134.0500 1.01134.0500
EM1.01134.0500CS 3496 USD
CA1.01134.0500 EM1.01134.0500
Ammonium carbamate, EMSURE® for analysis, Supelco®
Ammonium carbamate
Formula: H₂NCO₂NH₄
MW: 78.07 g/mol
MDL Number: MFCD00013010
CAS Number: 1111-78-0
EINECS: 214-185-2

Order Now

Specification Test Results

Solubility 790 g/l (20 °C)
Melting Point 152 °C (closed capillary tube)
Molar Mass 78.07 g/mol
Bulk Density 750 kg/m3
Vapor Pressure 82 hPa (20 °C)
Density 1.6 g/cm3 (20 °C)
pH 10 (100 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR