Barium diphenylamine-4-sulfonate redox indicator, Millipore®

Supplier: MilliporeSigma
Warning

Synonyms: Diphenylamine-4-sulphonic acid barium saltDiphenylamine-4-sulfonic acid barium saltN-Phenylsulfanilic acid barium saltN-Phenylsulphanilic acid barium salt

1.00255.0005 1.00255.0005
EM1.00255.0005CS 13885 USD
EM1.00255.0005 CA1.00255.0005
Barium diphenylamine-4-sulfonate redox indicator, Millipore®
Barium diphenylamine-4-sulphonate
Formula: C₂₄H₂₀BaN₂O₆S₂
MW: 633.89 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007497
CAS Number: 6211-24-1
EINECS: 228-278-0
UN: 1564
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/76/37/2297637.jpg

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 633.88 g/mol
Bulk Density 220 kg/m3
pH 5.9 (10 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic Materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR