Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3Prime Personal and Mid-Size Thermal Cyclers, Techne®

Supplier: Cole-Parmer
93945-01 93945-11 93945-05 93945-03 93945-09 93945-07
Techne have completely upgraded their established thermal cyclers to become the most feature rich range of instruments on the market

3PRIMEBASE key features include colour touchscreen user interface for fast program setup. Small space-saving footprint. Block options; 24 x 0.2ml (compatible with 8 tube strips) and 18 x 0.5ml. Fast ramp rate up to 3.0°C/sec. Data transfer via USB.

The 3PrimeX delivers all the features of the 3Prime instrument but with an expanded sample capacity, accommodating 48 x 0.2ml microtubes, 30 x 0.5ml tubes plus half a 96 well plate in a horizontal format. Design flexibility allows the unit to be upgraded to included gradient cycling capability.

The Prime range of thermal cyclers provide an unrivalled user experience because each unit, from the personal 3Prime right up to the full size PrimeG instrument feature a full colour touchscreen and intuitive user interface. The Prime range of thermal cyclers are also designed to provide easy programming. Pre-installed templates allow the user to quickly create even the most complicated programmes via the intuitive touch interface. All Prime thermal cyclers have the ability to transfer programmes between instruments via USB or to your PC. The internal memory can hold 1000 programmes with unlimited storage available on USB data stick. The Techne Prime range of thermal cyclers are among the most reliable thermal cyclers on the market, backed by a 4 year warranty, the best in the industry.

Ordering information: Four-year manufacturer's warranty.




 

ORDER

Expand All / Collapse All
Description Sample Capacity Block Format Supplier No. VWR Catalog Number Unit Price Quantity
3Prime Thermal Cycler 24 x 0.2 mL - 93945-01 89235-790 Each Retrieving
3Prime Thermal Cycler 18 x 0.5 mL - 93945-03 89235-792 Each Retrieving
3PrimeG Gradient Thermal Cycler 48 x 0.2 mL 48 x 0.2 mL 93945-09 89235-808 Each Retrieving
3PrimeG Gradient Thermal Cycler 30 x 0.5 mL 30 x 0.5 mL 93945-11 89235-810 Each Retrieving
3PrimeX Thermal Cycler (Can be Gradient Enabled with USB Upgrade) 48 x 0.2 mL - 93945-05 89235-798 Each Retrieving
3PrimeX Thermal Cycler (Can be Gradient Enabled with USB Upgrade) 30 x 0.5 mL - 93945-07 89235-800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Power 155W
Dimensions 350L x 210W x 180H mm
Weight 6 Kg
Temperature Accuracy ± 0.25°C at 55°C
Electrical 100-230V, 50-60Hz, 155W
Block Temperature Range 10°C to 100°C
Peltier Elements per Block 2 for 3Prime and 4 for 3PrimeX
Max. Heating Rate 3.0°C per second
Block Uniformity at 55 °C \<±0.3°C

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
3PrimeX block, 30 x 0.5 mL 93945-43 89235-804 Each Retrieving
USB Gradient Upgrade for 3PrimeX 3PRIMEXUSB 89235-806 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty