Plug Stopper, 17 mm, Globe Scientific
Supplier: Globe Scientific
116160
89497-862PK 77.78 USD
89497-862
Plug Stopper, 17 mm, Globe Scientific
Closures Test/Sample Tube Closures
This white polyethylene plug cap provides exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes.