Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Polystyrene and Polypropylene Test Tubes, 16 x 125 mm, Globe Scientific
Supplier: Globe Scientific
110115 110125
89497-778CS 188.17 USD InStock
89497-778 89497-780
Polystyrene and Polypropylene Test Tubes, 16 x 125 mm, Globe Scientific
Tubes Test Tubes
These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.
Polystyrene tubes offer a spin rate of 1500rpm. Polypropylene tubes offer a spin rate of 3000rpm.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty