Would you like to log in again?
CE Series Ovens, SHEL LAB
Supplier: Sheldon Manufacturing
SMO5 9960517 8070536 9960516 8070535 SGO3 8070531 SGO5-2 8070534 8070530 SGO3-2 SMO3 8070533 SMO5-2 SGO5 SMO3-2 8070532 8070537
89409-456EA 4204.12 USD InStock
89409-456 10356-018 10356-024 10456-412 10456-418 89409-452 10456-440 89409-462 10356-020 10356-042 89409-454 89409-448 10456-444 89409-458 89409-460 89409-450 10356-046 10456-422
CE Series Ovens, SHEL LAB
Ovens Standard Ovens
The CE Oven Series has triple wall construction and easy-to-read microprocessor controls
The forced air ovens feature a 99 hr 59 min digital timer that has an independent control, so use is optional. Once the timer expires, the heating elements turn off while the blower continues running to cool the samples inside the oven.
The gravity convection ovens provide a degree of uniformity comparable to most forced air ovens on the market. The 99 min 59 hr digital timer has an independent control so its use is optional. Once the timer expires the power is cut to only the heating elements.
Whatever the application, the forced air ovens and gravity convection ovens provide a degree of uniformity and the peace of mind expected from the SHEL LAB brand.
Users can adjust the air exhaust from the chamber without being near the heat source. Even at operation temperatures of 225°C, the outer skin of this unit meets all CE requirements, so workspaces remain cool
Certifications: All of the SHEL LAB CE series ovens are CE approved.
Ordering information: 2-year limited warranty.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: Calibration Services (Single and Multiple Point- Conducted Prior to Shipment at Sheldon Manufacturing. Not on Customer Site)–We offer ‘Calibration Service’ where we calibrate the temperature to a customer stated set point. The test configuration uses multi-point thermocouples (9 probes minimum) usually with 4 run minimum stable run times. When a customer wants multiple temperatures, we can test these parameters, but the unit can be calibrated only to one set point at a time. We will require the final number at which the unit will be calibrated when it ships. The documentation will show data for all requested calibration set points. Standard documents are Certificate of Calibration and graphs (shipped with the unit and .pdf version for email if needed).
Factory Acceptance Test (FAT, Conducted Prior to Shipping at Sheldon Manufacturing. Not on Customer Site )–The FAT is a customized test procedure for different types of systems and the tests are executed at the factory. The actual test details and elements are specified by the customer and agreed to by the company so that the test protocols are mutually specified. Prior to delivery, customers want to make sure that equipment operates as intended without disturbances occurring on site. In achieving this, customers are therefore able to verify that all specifications and contractual requirements have been met. In some cases, the customer may want to witness the testing as is the case with FAT. This is a custom quoted service for each type of test needed (i.e. CO2, temp, RH, load configuration, etc.).
Certificate of Conformity (Conducted Prior to Shipment at Sheldon Manufacturing. Not on Customer Site )–This is a serial number specific testing option which is done to verify the unit will perform to and meet all stated specifications. This includes Hipot, Ground Bond, Current draw for max settings, and calibration (when applicable). Testing is designed in such a way as to verify all operational aspects, but we may not test each specific attribute (for example, the unit max current draw and controller operation will tell us the unit is capable of heating to max temperature without actually heating to the stated max temperature). Standard documents are Certificate of Conformity and QA check list (scanned and kept on file). Additional Offering is the QA Check Sheet; these are scanned on unit completion, and can be provided “On Request-FOC”. These documents are kept a minimum of 7 years from date of manufacture.
Factory Acceptance Test (Conducted Prior to Shipment at Sheldon Manufacturing. Not on Customer Site )–The FAT is a customized test procedure for different types of systems and the tests are executed at the factory. The actual test details and elements are specified by the customer and agreed to by the company so that the test protocols are mutually specified. Prior to delivery, customers want to make sure that equipment operates as intended without disturbances occurring on site. In achieving this, customers are therefore able to verify that all specifications and contractual requirements have been met. In some cases, the customer may want to witness the testing as is the case with FAT. This is a custom quoted service for each type of test needed (i.e. CO2, temp, RH, load configuration, etc.).
Extended Warranty–The Extended Warranty Plan extends the equipment’s original warranty for an additional 1 year parts and labor (US and Canada only). This plan provides unlimited labor, parts, and travel, along with priority phone and e-mail support. The Extended Warranty option is available only at the time of the original equipment order. It is not available once the unit has been delivered.
Note: All service options are only valid at point of sale. Once you order the services for the corresponding products, Sheldon will mail you a work order documentation and in the meanwhile will hold the delivery of your products. Customers are required to fill the work order with your requirements such as calibration temperature point. After accomplish the work order, customers can either mail it back or Email scanned pdf to Sheldon (Email address will be provided in the work order). After receiving, Sheldon will conduct the services based on the work order.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty