Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Corning® PureCoat™ ECM Mimetic Cultureware, 24-Well Plates, Corning®
Supplier: CORNING DISCOVERY LABWARE PHASE II
356241 356271
89237-472CS 297.98 USD InStock
89237-472 89237-480
Corning® PureCoat™ ECM Mimetic Cultureware, 24-Well Plates, Corning®
Microplates Cell Culture Plates
Corning® PureCoat™ ECM Mimetic Cultureware is coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent alternative to complex extracellular peptides.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty