Would you like to log in again?
Personna® 0.009" Single Edge Razor Blades, Accutec Blades
Supplier: Accutec Blades
Personna®
94-0491
89427-046CS 929.33 USD InStock
89427-046
Personna® 0.009" Single Edge Razor Blades, Accutec Blades
Razors and Blades
Personna® Single Edge blades provide a product solution for virtually any need
Blades are degreased with a steel back and are supplied unwrapped. When productivity, cleanliness, and performance matter, choose Personna®.
They are designed for use in cleanrooms, medical device manufacturing, or other applications.
Ordering information: Blades are packaged 115 blades per clamshell container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty