Disposable Funnels, Globe Scientific
Supplier: Globe Scientific
8301 8302 8304 8303
60820-116CS 55.25 USD InStock
60820-116 60820-106 60820-110 60820-108
Disposable Funnels, Globe Scientific
Funnels Filtration Funnels Filter Funnels
These economical disposable funnels are available in either transparent polystyrene or translucent polypropylene
Available in either 55mm or 65mm widths.
They are lightweight, have reinforced rims and inside fluting. They are tapered at a 60° angle.