Would you like to log in again?
Snap Cap for 10 mm Tubes, Globe Scientific
Supplier: Globe Scientific
113139C 113139L 113139G 113139Y 113139R 113139B 113139A
89206-940BG 65.6 USD InStock
89206-940 89206-948 89206-946 89206-952 89206-950 89206-944 89206-942
Snap Cap for 10 mm Tubes, Globe Scientific
Closures Test/Sample Tube Closures
An effective method to recap blood drawing tubes, glass test tubes, and plastic culture tubes
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
The snap caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation, and refrigeration procedures. Available in a variety of bright and attractive colors for sample identification purposes.
Ordering information: Snap caps are supplied in a pack of 1000.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty