Immuno™ Immunodiffusion (RID) Plates, MP Biomedicals
Supplier: MP Biomedicals
ImmunO™
08421651
IC421651EA 238.92 USD
IC421651
Immuno™ Immunodiffusion (RID) Plates, MP Biomedicals
Microplates Immunology Microplates and Strips
Immunodiffusion Plates are used in Radial Immunodiffusion Technique.
  • Immunodiffusion Plates are polystyrene with close-fitting covers to minimize gel dehydration
  • Plate bottoms are recessed 7 mm to prevent scratching on the bench surfaces
  • The plate is hydrophilic for easy gel pouring and better adherence to the plate
Immunodiffusion Plates are used in Radial Immunodiffusion Technique which determine the quantity of an antigen by measuring the diameters of circles of precipitin complexes surrounding samples of the antigen that mark the boundary between the antigen and an antibody suspended in a medium.