Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
pMAL™-c5X Vector, New England Biolabs
Supplier: New England Biolabs (NEB)
N8108S N8108S
101641-058EA 141.45 USD InStock
CA101641-058 101641-058
pMAL™-c5X Vector, New England Biolabs
Vectors
The vector pMAL-c5X is designed to produce maltose-binding protein (MBP) fusions, where the protein of interest can be cleaved from MBP with the specific protease Factor Xa
MBP fusions made with this vector are expressed cytoplasmically. The MBP has been engineered for tighter binding to amylose resin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty