Would you like to log in again?
pMAL™ Protein Fusion and Purification System, New England Biolabs
Supplier: New England Biolabs (NEB)
E8200S
In the pMAL Protein Fusion and Purification System, the cloned gene is inserted into a pMAL vector downstream from the malE gene, which encodes maltose-binding protein (MBP).
  • Reliable expression
  • Substantial yields (up to 100 mg/L) in more than 75% of the cases tested
  • Expression in either the cytoplasm or periplasm
  • Periplasmic expression enhances folding of proteins with disulfide bonds
  • Fusion to MBP has been shown to enhance the solubility of proteins expressed in E. coli
  • Gentle elution with maltose
  • No detergents or harsh denaturants
This results in the expression of an MBP-fusion protein. The technique uses the strong Ptac promoter and the translation initiation signals of MBP to express large amounts of the fusion protein. The fusion protein is then purified by a one-step affinity purification specific for MBP. The system uses the pMAL vectors which include a sequence coding for the recognition site of a specific protease adjacent to the multiple cloning site. This allows the protein of interest to be cleaved from MBP after purification, without adding any vector-derived residues to the protein. For this purpose, the polylinker includes a restriction site superimposed on the sequence coding for the site of the specific protease. This is where the gene of interest is inserted. Expression from the pMAL vectors yields up to 100 mg fusion protein from a liter of culture. In most cases, the expressed protein is soluble, as fusion to MBP has been proven to enhance the solubility of proteins expressed in E. coli. While no expression system works with every cloned gene, the pMAL Protein Fusion and Purification System gives substantial yields of protein in more than 75% of the cases tested.
Ordering information: Reagents supplied with this product include pMAL-c5X Vector, pMAL-p5X Vector, Factor Xa Protease, E. coli ER2523 (NEB Express) (Glycerol Stock), Amylose Resin, MBP5 Protein, MBP5-paramyosin-Δsal, and Anti-MBP Monoclonal Antibody.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
pMAL Protein Fusion and Purification System E8200S 101640-924 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Small Business Enterprise
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty