Oligo (dT)25-Cellulose, New England Biolabs
S1408S
101228-118EA 139.4 USD
101228-118
Oligo (dT)25-Cellulose, New England Biolabs
An affinity matrix used for the isolation of mRNA containing polyadenylic (poly A) regions
This matrix consists of oligo (dT)25 covlaently coupled to a cross-linked cellulose bead. This product is guaranteed for twenty-four months when stored at 4°C in the supplied storage buffer.