Goat Anti-Mouse IgG Magnetic Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
S1431S S1431S
101228-122EA 192.7 USD InStock
CA101228-122 101228-122
Goat Anti-Mouse IgG Magnetic Beads, New England Biolabs
Beads Magnetic Beads
An affinity matrix for the small-scale immunomagnetic separation and purification of mouse IgGs
Anti-Mouse IgG is covalently coupled to a nonporous paramagnetic particle. This secondary antibody binds the heavy chain of mouse IgG and is suitable for immunoassays that employ a mouse IgG primary monoclonal antibody. Cell separations and sorting can be accomplished using a mouse IgG antibody to defined cell surface antigens.