Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
alpha 1-2 Fucosidase, New England Biolabs
Supplier: New England Biolabs (NEB)
P0724S P0724L P0724L P0724S
101446-532EA 533 USD InStock
CA101446-534 101446-532 CA101446-532 101446-534
alpha 1-2 Fucosidase, New England Biolabs
Enzymes
α1-2 Fucosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of linear α1-2 linked L-fucopyranosyl residues from oligosaccharides
In this case, a linear substrate is defined as having no branching on the adjacent residue.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty