Would you like to log in again?
VACUUBRAND® CVC300 Vacuum Controller and VACUU•BUS® Accessories, BrandTech
Supplier: BRANDTECH SCIENTIFIC INC.
612552 674217 636163 674220 674291 20683160 636657
89178-168EA 96.26 USD InStock
89178-168 89178-184 89178-170 89178-186 89178-188 89080-226 89178-174
VACUUBRAND® CVC300 Vacuum Controller and VACUU•BUS® Accessories, BrandTech
Pumps Vacuum Controllers
The VACUUBRAND® CVC3000 Vacuum Controller is an intuitive, user-friendly controller for the rough vacuum range
Featuring an integrate capacitive vacuum transducer and solenoid vent valve, and typically matched to a VVB6C solenoid (below) and a diaphragm vacuum pump, the CVC3000 will provide two-point control for a variety of laboratory tasks. It is also capable of executing complex multi-step programs, either programmed into the controller, or in conjunction with a PC and RS232 null modem cable (not included). It is also capable of management of a VACUULAN® vacuum local area network. With additional accessories the CVC3000 is capable of controlling rotary vane vacuum pumps in the fine vacuum range. Using the CVC3000 is simple, with menu-driven software and a jog-wheel interface, in fourteen different languages – making it an excellent choice for multi-ethnic labs. Two VACUUBUS ports allow the interface of accessories below for additional capabilities.
Solenoid Isolation Valve; the VVB6C chemistry isolation valve allows the use of VACUUBRAND® controllers to provide two-point control of most diaphragm pump vacuum applications. Fluoropolymer design provides maximum chemical compatibility. Transducers; the VSK3000 (capacitive) and VSP3000 (Pirani) vacuum transducers allow measurement from atmosphere to 0.1mbar/Torr/hPa or 10-3 mbar/Torr/hPa respectively, and integrate with both controllers and vacuum gauges with the VACUUBUS system. Both transducers have alumina ceramic wetted parts for corrosion resistance as well as shock resistance. Solenoid Vent Valve; the VBMB vent valve works with both controllers and gauges to allow remote venting of an application from the panel. Cooling Water Valve; the VVKWB valve integrates with the CVC3000 of a PC-system to turn on and off the cooling water to the emissions condenser as needed, allowing the efficient capture of solvent, while reducing water usage.
VACUUBUS Extension Cable; two meter cable allows a greater distance between controller and application, transducer and display or other options. VACUUBUS Y-Cable: Analogous to a USB hub, this “splitter” cable expands the number of VACUUBUS ports, for maximum flexibility.
Certifications: cTUVus (Model 89080-226 only).
Accessories information: VACUUBUS® accessories; in addition to fitting the CVC3000 controller, VACUUBUS® accessories fit the VNC2, the DCP3000 gauge and other VACUUBRAND® products. "Plug and play" compatibility automatically configures the controller or gauge for easy use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty