AccuTechTM 91-300, Ultra Clean Gloves, Ansell
Supplier: Ansell Healthcare
574127 574128 574129 574126 574124 574125 574123
79999-308CS 675.25 USD InStock
79999-308 79999-310 79999-312 79999-306 79999-302 79999-304 79999-300
AccuTechTM 91-300, Ultra Clean Gloves, Ansell
Gloves
Features and benefits of these ultra-clean gloves include an ergonomic design, hand-specific gloves, and exceptional dexterity.
  • Hand-specific
  • Exceptional dexterity
  • Roll-beaded cuffs
  • Ergonomic design
Cleanroom compatible, featuring a low particle glove with a low level of extractables. Powder-free. Made of 100% natural rubber, free of pigments or fillers. Roll-beaded cuffs make donning easier. Stays in position on the arm.
Ordering information: This product contains natural rubber latex which may cause allergic reactions in some individuals.