96 Well Clear Flat Bottom Poly-D-Lysine Coated Microplate, Aseptic, Corning®
Supplier: Corning
3842 3841 3843
89092-708CS 2396.39 USD InStock
89092-708 89092-706 89092-710
96 Well Clear Flat Bottom Poly-D-Lysine Coated Microplate, Aseptic, Corning®
Microplates Cell Culture Plates
Flat bottoms with 360µL total volume. Recommended working volumes of 75 to 200µL. Coated with poly-D-lysine for enhanced cell attachment. Nonreversable lids with condensation rings to reduce contamination. Sterile (aseptic assembly) and certified nonpyrogenic. Individual alphanumeric codes for well identification. Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescence and enabling readings down to 340nm. Bulk packed 20 per bag.
White and black options have opaque walls to prevent well-to-well crosstalk. Optically clear flat well bottom permits direct microscopic viewing. Can be used for both top and bottom reading instruments. White walled plates enhance luminescent signals and have low background luminescence and fluorescence. White walled plates enhance luminescent signals and have low background luminescence and fluorescence. Black wall plates have low background fluorescence, minimal light scatter.
White and black options have opaque walls to prevent well-to-well crosstalk
Optically clear flat well bottom permits direct microscopic viewing. Can be used for both top and bottom reading instruments. White walled plates enhance luminescent signals and have low background luminescence and fluorescence. White walled plates enhance luminescent signals and have low background luminescence and fluorescence. Black wall plates have low background fluorescence, minimal light scatter.