Stopcock Replacement Plug, 1:5, Straight Bore, Fluoropolymer Resin, Glassco
Supplier: Glassco
300.470.03
89426-780EA 11.12 USD
89426-780
Stopcock Replacement Plug, 1:5, Straight Bore, Fluoropolymer Resin, Glassco
Stopcocks
Replacement kit is supplied complete with one fluoropolymer resin plug (1:5 taper), one teflon washer, one O-Ring, and one plug with PTFE handle
PTFE nut is also available.