GRAB-EEZ™ ESD Cleanroom Wipe Dispenser and Wipes, High-Tech Conversions
Supplier: HIGH-TECH CONVERSIONS
GE-NT1-712 GE-DISP1
10146-288CS 370.2 USD InStock
10146-288 89492-640
GRAB-EEZ™ ESD Cleanroom Wipe Dispenser and Wipes, High-Tech Conversions
Dispensers Wipe Dispensers
The Grab-EEZ™ is an ESD safe plastic cleanroom wipe dispenser
NOVA-TECH™ 1000 is a tough, abrasion resistant, absorbent, and virtually free of particle generation, non-woven cleanroom wipe. It is comprised of a synergistic blend of virgin polyester and cellulose that is hydroentangled into a uniform, super-tough fabric. These unique features give NOVA-TECH™ 1000 a high degree of absorbency and effectiveness during the process of general wiping. Wipes made with this fabric are converted in a class 100 cleanroom.
This dispenser can be placed on any countertop or mounted to the wall. Features specifically designed 7X12" c-folded wipes which are larger than the conventional 9X9" size. There is a viewing slot, so you can see when you are getting low on wipes. The wipe opening has been specially designed, so there is a low particulate released into the air and so it allows you to easily grab only one wipe with zero cross contamination. ISO Class 5(Class 100) compatible.
Ordering information: Unit comes with ESD Plastic wipe holder, ESD Cover, ESD Stand, Weight/Tamper, and Angle Adjustment & Lock. Wipes come 200 per pack/ 12 bags per case/ 60 cases per pallet.