VACUUBRAND® VARIO® Vacuum Pumps and PC Systems, BrandTech
Supplier: BrandTech
20738403 20732403 20733503 20736503 20737503
89080-224EA 18594.39 USD
89080-224 89080-220 89178-224 89080-208 89080-216
VACUUBRAND® VARIO® Vacuum Pumps and PC Systems, BrandTech
VARIO® Chemistry-design vacuum pumps and systems combine fluoropolymer diaphragm vacuum pumps with speed controlled motor, pressure transducers to measure pressure change over time, and sophisticated software
Deepest Vacuum in a Chemistry Design Diaphragm Pump. The PC3003 has an ultimate vacuum of 4.5 x 10-1 Torr, the deepest vacuum available in a chemistry design diaphragm vacuum pump.
Chemistry design. VARIO® chemistry design diaphragm pumps have a full fluoropolymer flowpath for high resistance to solvent and acid vapors. They also feature high flow rates at working vacuum for more work performed. An integrated gas ballast allows continuous purge of internally condensed vapors. "Chemistry design" means no oil to change or dispose of – ever.
This innovative set of features allows VARIO® pumps to perform tasks impossible with other technologies: Find and follow boiling point curves with virtually no bumping, all without pre-programming; single-point "zero-hysteresis" vacuum control; save up to ten multi-step vacuum programs...and more. Because the speed controlled motor only pumps as much as necessary, they are virtually silent in operation, and diaphragm life is well in excess of the typical 15,000 operational hours of our pumps running full speed.