Would you like to log in again?
Chloride Test Method: Colorimetric With Test Strips (5 Reaction Zones) 500 - 1000 - 1500 - 2000 - ≥ 3000 mg/l Cl- MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10079.0001 1.10079.0001
CA1.10079.0001EA 79.04 USD InStock
CA1.10079.0001 EM1.10079.0001
Chloride Test Method: Colorimetric With Test Strips (5 Reaction Zones) 500 - 1000 - 1500 - 2000 - ≥ 3000 mg/l Cl- MQuant™, MilliporeSigma
Environmental Test Kits
The chloride test is suitable for measuring chloride ions in all natural waters
MilliporeSigma # 1.10079.0001 replaces and is identical to MilliporeSigma # 10079-1
Their concentration depends on geological factors and the general local situation.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty