Would you like to log in again?
Vacuum Filter Tube, with Inner Standard Taper Joints on Top and Bottom, Ace Glass Incorporated
Supplier: ACE GLASS, INC.
7827-01 7827-03 7827-05 7827-07
This borosilicate glass Schlenk filter tube features an integral glass filter disc at the tube's mid-line, and inner joints at top and bottom.
ORDER
Expand All / Collapse All
Description [ST] Joint No. Porosity Supplier No. VWR Catalog Number Unit Price Quantity
Vacuum Filter Tube with Inner Standard Taper Joints on Top and Bottom 14/20 25–50 µm 7827-01 89428-000 Each Retrieving
Product available on GSA Advantage® Small Business Enterprise
Vacuum Filter Tube with Inner Standard Taper Joints on Top and Bottom 14/20 10–20 µm 7827-03 89428-002 Each Retrieving
Product available on GSA Advantage® Small Business Enterprise
Vacuum Filter Tube with Inner Standard Taper Joints on Top and Bottom 24/40 10–20 µm 7827-05 89428-004 Each Retrieving
Product available on GSA Advantage® Small Business Enterprise
Vacuum Filter Tube with Inner Standard Taper Joints on Top and Bottom 24/40 25–50 µm 7827-07 89428-006 Each Retrieving
Product available on GSA Advantage® Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty