Ward's® External Heart Rate Sensor Ear-Clip
Ward's
LD12-HR
470210-534EA 65.99 USD
470210-534
Ward's® External Heart Rate Sensor Ear-Clip
Expand the probeware versatility of your Datahub.
  • Easy to use
  • Accurate and precise
Measure, record, and monitor heart rate using any Ward's DataHub model with the addition of this external sensor. Just plug into the universal sensor adapter. Includes an ear clip adapter.