Ward's® External Respiration Sensor
Ward's
LD12-SPR
470210-532EA 87.99 USD
470210-532
Ward's® External Respiration Sensor
Expand the probeware versatility of your Datahub.
  • Easy to use
  • Accurate and precise
Record and measure the rate of respiration using any Ward's DataHub model with the addition of this external sensor. Just plug into the universal sensor adapter.