Multi-Purpose Test Tubes, Stockwell Scientific
Supplier: Stockwell Scientific
8562AM 8562P 8550 8552 8553 8561 8562Y 8562L 8560 8572 8562O 8562 8594 8562G 8562B 8554
14224-252CS 186.42 USD
14224-252 60820-462 60820-410 60820-432 60820-400 60820-412 60820-456 60820-402 60820-414 60820-404 60820-448 89209-886 60820-416 60820-406 60820-408 89050-200
Multi-Purpose Test Tubes, Stockwell Scientific
These multi-purpose test tubes are precision molded from premium-grade plastics and ideal for routine laboratory analysis and procedures.
  • Tubes are avalable in several sizes and colors in two material formats
  • The polystyrene tubes are clear like glass, and the polypropylene tubes are chemically resistant
Maximum use temperature for polystyrene is 70 °C, and polypropylene is 135 °C. The brittleness temperature for polystyrene is 20 °C, and for polypropytene it is 0 °C.
Ordering information: Tubes are supplied without caps.