Vortex 4 Basic and Digital Vortexers, IKA® Works
4050100 4049600
89495-290EA 860.09 USD
89495-290 89495-292
Vortex 4 Basic and Digital Vortexers, IKA® Works
Compact, universal shakers are suitable for shaking tasks with small vessels and microtiter plates.
Vortex 4 Basic and Digital Vortexers accommodate a wide range of attachments, and provide attachment detection. Units are stable in all speed ranges and feature a sturdy zinc die cast casing. The Basic unit features continuous and touch operation, while the Digital unit features timer, continuous, and touch operation. Two operating modes are standard. In Mode A (safe mode with attachment detection) the maximum speed of 3000rpm is only reached with the standard attachment in touch mode. When using other attachments the speed is limited to 1300rpm. In Mode B (without attachment detection) a speed of 3000rpm is possible with all attachments.
Units meet IP 40 protection class according to DIN EN 60529.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration at www.ika.net/register). Accessories are sold separately.