CO2 Water Jacket Incubator, SHEL LAB
SCO10W SCO2W SCO5W
89409-288EA 5922.9 USD
89409-288 89409-290 89409-292
CO2 Water Jacket Incubator, SHEL LAB
The 3500 series of CO2 Incubators offer dependable Infrared (IR) CO2 Sensor control and are ideal for sensitive tissue and cell culture applications
For the fastest CO2 recover and most stable performance, this series features IR sensors. Extensive use of copper in the CO2 sample port, heated CO2 feed line, housing of the patented HEPA filtration system, and humidity pan with a copper SL decontamination token, adds reassurance that foreign microbes will not affect test results. Cleanup is a breeze with the all stainless steel chamber and autoclavable door gasket. Optional copper shelves are available for even more contamination control. A "Bacteriostatic" copper cage to trap particulate matter and reduce potential for chamber contamination. This filter removes 99.97% of all airborne microbes and isolated microes 0.3 micons or larger.
These incubators control three essential variables related to replicating the mammalian environment: Stable CO2 Level, Controlled Temperature, and Relative Humidity.
They provide the benefits of contamination control and uncompromising temperature uniformity for even the most demanding incubations. Precision is easily maintianed with push-button calibration of both temperature and CO2, and audio/visual alarms that signal high/low temperature and CO2 conditions. Modular controls and backup systems ensure confidence for incubating valuable samples, providing the dependable assurance you expect from a SHEL LAB incubator.
Ordering information: 5 year parts (labor included in the US). 7 year IR sensor. Lifetime water jacket chamber.