JACKSON SAFETY® V50 EPIC™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: KIMBERLY CLARK CORP
JACKSON SAFETY®
33345 33347 35327
89427-040CS 403.62 USD InStock
89427-040 89427-042 89427-044
JACKSON SAFETY® V50 EPIC™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Lightweight design, soft nosepiece and soft, flexible, non-slip temples for comfort
Soft padded liner provides complete seal to face to protect from dust and debris. Top and bottom direct ventilation combined with VISICLEAR 2™ anti-fog coating provides exceptional protection against fogging (two times the "N" anti-fog standard according to EN166). Headstrap can be changed to temple bars with an easy click of a button. Base curve of 8.0. Glasses provide 99.9% UV protection against harmful UV rays. Meets ANSI Z87.1+ 2010 Standards. CSA Certified.