Jackson Safety® G60 Level 3 Knuckle Coated, High-Visibility Cut Resistant Gloves with Dyneema® Fiber, Kimberly-Clark
38647
89426-994CS 307.92 USD
89426-994
Jackson Safety® G60 Level 3 Knuckle Coated, High-Visibility Cut Resistant Gloves with Dyneema® Fiber, Kimberly-Clark
Add cut resistance to your worker's gloves for tasks that require protection against hand lacerations with the new JACKSON SAFETY® G60 Level 3 Knuckle coated, high-visibility cut resistant gloves with Dyneema® fiber coated with polyurethane.
  • Dyneema® yarn with high-visibility nylon thread
  • Lycra® fiber provides cut resistance plus enhanced breathability and comfort
  • Dark coating hides dirt for longer wear
An ideal choice for working environments that pose a risk of minor cuts and abrasions. Black polyurethane coating provides excellent dry and oil grip and great dexterity for improved productivity. Typical uses include metal fabrication, glass handling, automotive assembly, stamping, and assembly.
Certifications: Meets ANSI Level 2/EN388 Level 3 cut resistance and EN 420 standards.