Cryotome FSE Cryostat, Thermo Scientific
A78900004H A78900004L A78900004K
89428-776EA 31471.65 USD InStock
89428-776 89428-780 89428-778
Cryotome FSE Cryostat, Thermo Scientific
Microtomes and Microtome Assemblies Microtomes
The Thermo Scientific Cryotome FSE is a safe and reliable cryostat designed for operator comfort, and is also adaptable to multiple users and samples. Color-coded levers and graphics guide knifeholder adjustments and a simple, intuitive, multi-lingual touch screen display provides easy instrument operation. The angle of the clamp plate enables easy specimen pick-up.
The Cryotome FSE provides fast specimen turn-around time through the use of a cryobar with adjustable temperature controls and a dedicated cryobar refrigeration system that rapidly cools the specimen to help prevent freezing the artifact
The microtome is located outside the cryochamber for faster temperature adjustment and ease of decontamination and cleaning. An automated fumigation cycle disinfects the cryochamber, and a dual compressor system ensures specimen integrity by keeping specimens frozen if the cryobar fails. Multi-level security code protection and the lockable heated window prevent unauthorized access and protocol alterations.
A positive locking flywheel prevents injury while working around the blade and specimen. The three-level adjustable footrest helps reduce fatigue while sitting or standing, and rounded exterior edges and a raised cutting area help prevent back strain while sectioning. The units use CFC-free refrigerants and foam insulation.
The Cryotome FSE cools rapidly from ambient to set temperature in as little as two hours. The "quickheated" specimen release feature allows easy removal of frozen specimen from the cryocassette. The Cryotome FSE features an orienting head with specimen holder temperature control that allows for enhanced sectioning of a variety of tissue types.
Certifications: CE marked. CSA certified. UL listed (Standard 3101).
Ordering information: Blade holder is not included and must be specified at time of order. Cryotomes are supplied with a specimen head, cryocassettes of various sizes and styles, cryobar with cryocassette identifier and plastic cover, heat extractors, specimen and tool storage shelves, debris tray and brush, and operator manual. Additional accessories are available. Contact your VWR® representative for more information.