VWR® Laboratory Refrigerators with Solid Doors, 1 to 10°C
SCLP-3004 SCLP-2004
97058-846EA 6265.81 USD
97058-846 97055-698
VWR® Laboratory Refrigerators with Solid Doors, 1 to 10°C
General-purpose, upright laboratory refrigerators feature a microprocessor temperature controller with digital display that allows precise temperature adjustment, maintenance, and recovery.
  • General-purpose use
  • User-adjustable alarms
  • Extensive security package
  • Precise temperature adjustment
Units also feature an extensive temperature security package, including user-adjustable audible and visual alarms for high and low temperatures and power failures, battery backup, a temporary muting option with intermittent chirp reminder, sample simulated digital temperature display, and sensor error warning. Remote alarm contacts with adjustable delay simplify connection to central alarm systems and phone dialers. A solution bottle is installed within each unit to provide an accurate indication of sample temperatures. Refrigerators are cycle preset to maintain an average temperature of 4°C (39.2°F). Supplied with 10.2 cm (4") swivel casters. Shelves included.
Certifications: UL and C-UL listed.
Ordering information: Refrigerators are supplied with a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.