Linear Shaking Water Bath, 18 L, Grant Instruments
GLSAQP18US
97055-830EA 4411.8 USD InStock
97055-830
Linear Shaking Water Bath, 18 L, Grant Instruments
Baths Shaking Water Baths
High quality, high precision linear shaking water bath from Grant Instruments
The 18 liter high grade stainless steel tank has a durable polished finish. It features two programmable temperature and shaking presets - allowing frequently used temperatures and shaking speeds to be quickly selected. The digital PID temperature control ensures quick heat-up and precision control throughout the temperature range. The external steel case is powder coated for maximum chemical and abrasion resistance. The user can adjust an independent over-temperature sample protection function which protects samples from possibly over-heating. It is very safe to operate for extended periods due to a fixed thermal cut-out which protects the user if the bath is accidentally run without water or in the very unlikely event of failure of both control systems. Its operation is very accurate and the user can calibrate the bath at either a single or dual-point temperatures for optimum accuracy.
The GLS key features include ambient +5ºC to 99ºC operation; stability ± 0.1ºC; drain tap allows convenient emptying; raised feet allows lifting while holding base of tank; heater mat and sensor bonded to underside of tank, optimises temperature uniformity, workspace and is easy to clean; Grant non-drip polycarbonate lid, included as standard, improves performance, limits evaporation, and conserves energy; countdown timer with audible buzzer for accurate reaction timing; front panel lock-out disables front panel controls and prevents settings being accidentally changed; clear, wide-angle viewing digital LED display with indication of heating state –'heating-up', 'cooling down', or 'maintaining temperature'.
There are a wide range of accessories to provide exactly the right solution for your application, from basic through to sophisticated needs, each model represents excellent value for money.
High accuracy PID temperature control combined with a robust, high quality, linear shaking mechanism guarantees years of operation. The patented magnetically coupled shaking mechanism works smoothly, quietly and consistently even in demanding applications.
Certifications: CE Approved.
Ordering information: Supplied complete with instructions and power cord. Extended 3 year warrantee as standard. Please select a suitable shaking tray.