Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Economy Gravity Ovens, SHEL LAB
SGO4E SGO2E-2 SGO1E SGO6E SGO2E SGO4E-2 SGO6E-2 SGO1E-2
89409-444EA 1611.88 USD InStock
89409-444 89409-442 89409-436 89409-404 89409-440 89409-446 89409-406 89409-438
Economy Gravity Ovens, SHEL LAB
Ovens Standard Ovens
The SHEL LAB economy ovens feature a gravity convection design for efficient heating
Gravity Convection units are preferable for applications with lightweight samples (powders, lightweight films and granular materials) because they do not have an internal fan that will blow these samples around within the chamber.
Features: 3" adjustable exhaust port, adjustable stainless steel shelves, independent overtemperature safety, high temperature silicone door goblet, thermometer and clip supplied. Oven Applications: batch testing, aging tests, sterilizing, electronic burn-in, conditioning, preheating process, components processing, stability testing, glassware drying, asphalt testing.
The double wall insulation and 2 inches of industrial fiberglass insulation help to minimize heat loss. The low watt density heater keeps temperatures uniform and keep your energy consumption down
Ordering information: 2 Year Limited Warranty.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty