Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Shaking Incubators, SHEL LAB
Supplier: Sheldon Manufacturing
SSI5R-HS SSI5 SSI5R
89409-372EA 18408.66 USD InStock
89409-372 89409-368 89409-370
Shaking Incubators, SHEL LAB
Shakers and Mixers Shaking Incubators
The Shelf Lab Incubator Shakers are fine examples of our constant thinking
The SI6 and SI6R feature our unique counter balance weighting system that is adjustable to accommodate off-center loads, varying capacities and stroke lengths, which in turn allows for smoother operation. An adjustable orbit allows the user to change the rotational stroke from 0.5Î (vigorous shaking) to 1Î (gentle shaking) to accommodate different types of cells and to provide maximum oxygen transfer. The SI6 has a small footprint and its chamber provides shelf space for static incubation during shaking. The SI6R refrigerates via 1/6-horsepower motorized compressor (produced without CFCs/HCFCs).
All major functions–temperature, RPM and time–have alarms that alert the user to deviations from set parameters. Overtemperature protection is provided via a safety thermostat. Digital keypad operation offers the ability to calibrate the temperature controller to a reference thermometer. To better support the load over years of constant use, we utilize four load bearing positions for optimal weight distribution.
These incubators will provide a controlled environment for continuous growth of biological organisms. Market applications include microbiology, tissue culture, cell culture, and bacteriology.
Ordering information: 2 Year Limited Warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty