Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
VWR® Gravity Convection Incubator, Basic, 120V
VWR®
VWIG-50.3 VWIG-155.3 VWIG-32.3 VWIG-105.3
414005-130EA 2578.1 USD InStock
414005-130 414005-134 414005-128 414005-132
VWR® Gravity Convection Incubator, Basic, 120V
Incubators Standard Incubators
VWR® Basic Gravity Convection General Incubators are designed for convenient and safe use in many applications, such as Animal & Plant Cell Culture, Constant Temperature, and Germination Test.
  • Microprocessor control
  • Rotary knob interface
  • Temperature range: Ambient +8 to 65°C
  • Over-temperature & over-current protection
  • Sensor error detection
  • RS-232C interface for monitoring and controlling with PC (Protocol provided, no software)
All incubator units feature a 3.3 cm (1.3") stainless steel ventilation cap, two doors (one solid, one glass), high temperature grade foamed silicone rubber door gaskets, over-temperature and over-current protection, and sensor error detection
Note: This unit is not designed for use with corrosive substance applications.
Units are constructed with a durable, powder-coated steel exterior, stainless steel interior, two stainless steel shelves, and glass wool insulation. A locking mode assists in preventing unintended temperature changes. Units are also equipped with an internal 110V outlet for auxillary equipment.
Ordering information: Units come with a two-year parts and labor warranty. Incubators include two shelves. 230V, 50/60Hz models are also available; contact your VWR sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
SERVICES
Services: VWRCATALYST™ offers customer on-site Calibration, Preventative Maintenance and validation services including IQ, OQ, and PQ protocol development and execution services (US only).Please Call 888-793-2300 or visit www.vwr.com/equip_services for more details.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty