Would you like to log in again?
PIG® Brown Oil-Only Absorbent Mat Pad, New Pig
Supplier: NEW PIG CORP
MAT503 MAT502 MAT504
56620-046CS 138.94 USD InStock
56620-046 56620-058 56620-048
PIG® Brown Oil-Only Absorbent Mat Pad, New Pig
Spill Control Materials
Brown color blends in with most environments for both indoor and outdoor use; hides grime, so mat stays on the job longer
Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water. Floats to clean up spills on water. Can be wrung out and incinerated after use to reduce waste or for fuels blending. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Heavyweight construction is highly durable and absorbent for demanding tasks and large-volume cleanups. Mediumweight construction is versatile for general, everyday tasks and cleanups. Light-weight construction is economical for small tasks and cleanups.
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty