Would you like to log in again?
TIMP-1 Human, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Biotrak™
RPN2611 RPN2611
95019-110EA 998 USD InStock
95019-110 CA95019-110L
TIMP-1 Human, GE Healthcare
Assays Immunological Assays
  • Specific for total TIMP-1 free TIMP-1 and TIMP-1 complexed to MMPs
  • Measures total TIMP-1
  • Specific for human TIMP-1
  • Precise and accurate measurement
  • Ready to use substrate
  • Non-isotopic protocol
  • Same day protocol
Suitable for use with cell culture supernatant, serum, and plasma samples. The assay is based on a two site ELISA 'sandwich' format. Standards and samples are incubated in microplate wells pre-coated with anti-TIMP-1 antibody. Any TIMP-1 present will be bound to the wells, other components of the sample being removed by washing and aspiration. The TIMP-1 is detected by a peroxidase labeled antibody to TIMP-1. Any excess is removed by washing and aspiration. The amount of peroxidase bound to each well is determined by the addition of TMB 'ready to use' substrate. The reaction is stopped by addition of an acid solution, and the resultant colour read at 450nm in a microtitre plate spectrophotometer. The concentration of TIMP-1 in a sample is determined by interpolation from a standard curve. Each pack contains reagent for 96 determinations. This allows the construction of a standard curve plus the measurement of 42 samples in duplicate.
The assay recognises total human TIMP-1, ie. free TIMP-1 and that complexed with MMPs. The assay will fully cross-react with TIMP-1 in complexes with MMP-1, MMP-3, MMP-2, MMP-9. It does not cross-react with TIMP-2. It does not recognise TIMP-1 from other species. The cross-reactivity with MMP-1 and MMP-1/TIMP-1 complex for example was tested. The lack of interference of MMP-1 in the presence of the TIMP-1 standards was also tested.
Ordering information: Kit contents include Microplate: 12 x 8 well strips coated with anti-TIMP-1; Assay buffer 1: 10mL of phosphate buffer concentrate; Standard: 100ng lyophilized TIMP-1; Peroxidase conjugate: Lyophilized anti-TIMP-1 horseradish peroxidase; Wash buffer: 12.5mL phosphate buffer concentrate; TMB Substrate: 3,3’, 5,5’-Tetramethylbenzidine (TMB)/hydrogen peroxide.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty